Order Entry
Italy
ContactUsLinkComponent
VWR® Bioal Rack for Bio-Protection-Ser Container
VWR® Bioal Rack for Bio-Protection-Ser Container
Catalog # PBIB12443
Supplier:  VWR Collection
undefined
VWR® Bioal Rack for Bio-Protection-Ser Container
Catalog # PBIB12443
Supplier:  VWR Collection

Specifications

  • For tube Ø:
    61 mm
  • No. of Holes:
    8
  • Description:
    Bioal
  • W×D×H:
    310×140×70 mm
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

Anodised aluminium.

Rack for transport of urine sample containers within the Bio-Protection carrier.