Tube reader, PYROS Kinetix® Flex
Certificates
About this item
The PYROS Kinetix® Flex tube reader is an optical tube reader that runs both chromogenic and turbidimetric assays in compliance with the USP, EP and JP bacterial endotoxin test chapters.
- 32, 64 and 96 wells
- For the detection of endotoxins (detection limit: 0,001 EU/ml) to be used with - PYROCHROME® for the kinetic chromogenic LAL method, - PYROTELL®-T for the kinetic turbidimetric LAL method, - PYROTUBE®-K reaction tubes (7.9×75 mm) made of borosilicate glass
Delivery: All instruments inclusive: PYROS® EQS software with manual (21 CFR Part 11 Compliant), PYROS® EQS validation protocol package (IQ, OQ, PQ, SQ), tube rack.
Frequently Bought Together