Diethyl allylphosphonate 97%
:1067-87-4
:
:C₇H₁₅O₃P
:MFCD00015134
:178.1660 g/mol
:97 °C
:Ambient
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4325 to 1.4345 (20°C, 589 nm)
- GC:≥96.0 %
Frequently Bought Together