
(R)-(-)-6,6'-Dibromo-1,1'-bi-2-naphthol ≥98.0% (by HPLC)
Catalog # TCIAD2729-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 96.0 %
- Specific rotation [a]20/D:-44.0 to -50.0 deg (C=2,THF)
Specification Test Results
About this item
:
:444.12 g/mol
:GHS07
CAS Number:65283-60-5
Formula:C₂₀H₁₂Br₂O₂
MDL Number:MFCD00798290