Order Entry
Italy
ContactUsLinkComponent
 
 
5,5-Dibromo-2,2-dimethyl-1,3-dioxane-4,6-dione (5,5-Dibromomeldrum's acid) ≥98.0% (by titrimetric analysis)
 :  TCI
 
undefined
 
5,5-Dibromo-2,2-dimethyl-1,3-dioxane-4,6-dione (5,5-Dibromomeldrum's acid) ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Iodometric Titration):
    min. 98.0 %
  • Melting point:
    74.0 to 78.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

Frequently Bought Together