Order Entry
Italy
ContactUsLinkComponent
Diethyl-4-methoxyphenylphosphonate ≥97.0% (by GC)
Diethyl-4-methoxyphenylphosphonate ≥97.0% (by GC)
Catalog # TCIAM1152-10G
Supplier:  TCI
CAS Number:  
undefined
Diethyl-4-methoxyphenylphosphonate ≥97.0% (by GC)
Catalog # TCIAM1152-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :244.23 g/mol
EINECS:214-548-5
CAS Number:3762-33-2
Formula:C₁₁H₁₇O₄P
MDL Number:MFCD00015670