3,4-(Methylenedioxy)phenylacetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2861-28-1
:
Formula:C₉H₈O₄
MDL Number:MFCD00014576
:180.16 g/mol
EINECS:220-679-9
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:128.0 to 131.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together