Order Entry
Italy
ContactUsLinkComponent
 
3,4-(Methylenedioxy)phenylacetic acid 98.5+%
 
undefined
3,4-(Methylenedioxy)phenylacetic acid 98.5+%

 

  • Appearance:
    Beige to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    128 to 132 °C
  • Water:
    ≤0.5 % (K.F.)
  • Sulfated ash:
    ≤0.5 %
  • Titration with NaOH:
    ≥98.50 % (On dry substance)

 

 

Frequently Bought Together