
5-Nitro-1,3-benzodioxole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2620-44-2
:
Formula:C₇H₅NO₄
MDL Number:MFCD00005824
:167.12 g/mol
EINECS:220-055-6
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:146.0 to 150.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together