Order Entry
Italy
ContactUsLinkComponent
5,8-Dihydroxy-1,4-naphthoquinone ≥75.0% (by HPLC)
Irritant
5,8-Dihydroxy-1,4-naphthoquinone ≥75.0% (by HPLC)
Catalog # TCIAD2070-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
5,8-Dihydroxy-1,4-naphthoquinone ≥75.0% (by HPLC)
Catalog # TCIAD2070-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Dark red to Brown powder to crystaline
  • Purity (HPLC):
    min. 85.0 area%
  • Purity (Neutralization titration):
    min. 85.0 %

Specification Test Results

About this item

 :
 :190.16 g/mol
EINECS:207-495-4
 :GHS07
CAS Number:475-38-7
Formula:C₁₀H₆O₄
MDL Number:MFCD00001685