Order Entry
Italy
ContactUsLinkComponent
 
 
5-Hydroxy-1,4-naphthoquinone (juglone) ≥96.0% (by GC)
 :  TCI
 
undefined
 
5-Hydroxy-1,4-naphthoquinone (juglone) ≥96.0% (by GC)
 :  TCI

 

  • Appearance:
    Orange to Amber to Dark red powder to crystal
  • Purity (GC):
    min. 96.0 %

 

Frequently Bought Together