

2-Hydroxy-1,4-naphthoquinone 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:83-72-7
:
Formula:C₁₀H₆O₃
MDL Number:MFCD00001678
:174.16 g/mol
Merck Index:14,5393
Melting Pt:∼ 193 °C (dec.)
EINECS:201-496-3
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together