Order Entry
Italy
ContactUsLinkComponent
2-Hydroxy-1,4-naphthoquinone 99%
Irritant
Health
2-Hydroxy-1,4-naphthoquinone 99%
Certificates
undefined
Irritant
Health
2-Hydroxy-1,4-naphthoquinone 99%
Specification Test Results
  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥98.5 %
  • GC:
    ≥98.5 %