
1,2-Difluoro-4,5-dimethoxybenzene ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:203059-80-7
:
Formula:C₈H₈F₂O₂
MDL Number:MFCD00070789
:174.15 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to lump
- Purity (GC):min. 95.0 %
- Melting point:41.0 to 45.0 deg-C
Specification Test Results
Frequently Bought Together