
1,4-Dihydroxy-2-naphthoic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD2296-25G
Supplier: TCI
CAS Number:
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
About this item
:
:204.18 g/mol
EINECS:250-674-7
:GHS07
CAS Number:31519-22-9
Formula:C₁₁H₈O₄
MDL Number:MFCD00010370