
2-Chloro-5-iodobenzotrifluoride ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:260355-20-2
:
Formula:C₇H₃ClF₃I
MDL Number:MFCD09037801
:306.4514 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to lump
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together