Organic reference standard, 4-Nitrophenol 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL15590400AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:100-02-7
- Description:4-Nitrophenol 10 µg/ml in acetonitrile
- Formula:C₆H₅NO₃
- Description:4-Nitrophenol