1-(2-Thienyl)-1-propanone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:13679-75-9
:
Formula:C₇H₈OS
MDL Number:MFCD00005446
:140.20 g/mol
Boiling Pt:228°C
Flash Pt:101°C
EINECS:237-182-8
Specification Test Results
- Appearance:Colorless to Red to Green clear liquid to cloudy liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together