Order Entry
Italy
ContactUsLinkComponent
Traumatic acid (trans-2-dodecene-1,12-dioic acid) ≥90.0% (by GC)
Traumatic acid (trans-2-dodecene-1,12-dioic acid) ≥90.0% (by GC)
  TCIAD0029-100MG
 :  TCI
 :  
undefined
Traumatic acid (trans-2-dodecene-1,12-dioic acid) ≥90.0% (by GC)
  TCIAD0029-100MG
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Seq:
    2440
  • Environmentally Preferable:

 

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 90.0 %
  • Melting point:
    161.0 to 167.0 deg-C

 

 

 : trans-2-Dodecene-1,12-dioic acid, Traumatic acid
 :
 :14,09576
 :229-019-4
 :GHS07
 :6402-36-4
 :C₁₂H₂₀O₄
 :MFCD00004443