N-Acetyl-L-methionine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:65-82-7
:
Formula:C₇H₁₃NO₃S
MDL Number:MFCD00064441
:191.2 g/mol
Merck Index:14,00096
EINECS:200-617-7
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 98.0 ee%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:103.0 to 107.0 deg-C
Specification Test Results
Frequently Bought Together