Order Entry
Italy
ContactUsLinkComponent
 
2-Methyl-3-(methylthio)pyrazine ≥99.0% (by GC)
  TCIAM0685-1ML
 :  TCI
 :  
undefined
2-Methyl-3-(methylthio)pyrazine ≥99.0% (by GC)
  TCIAM0685-1ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

 

 

 :
 :140.21 g/mol
 :104°C
 :76°C
 :220-736-8
 :2882-20-4
 :C₆H₈N₂S
 :MFCD00014610