2-Ethyl-3-(methylthio)pyrazine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:72987-62-3
:
Formula:C₇H₁₀N₂S
MDL Number:MFCD00038027
EINECS:277-191-4
Specification Test Results
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together