Order Entry
Italy
ContactUsLinkComponent
 
 
2,3-Dicyano-5-methylpyrazine ≥99.0% (by GC)
 :  TCI
 
undefined
 
2,3-Dicyano-5-methylpyrazine ≥99.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    98.0 to 101.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

Frequently Bought Together