
2,3-Dicyanopyrazine ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:13481-25-9
:
Formula:C₆H₂N₄
MDL Number:MFCD00059771
:130.1069 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:131.0 to 135.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together