
1-Acetyl-2-imidazolidinone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5391-39-9
:
Formula:C₅H₈N₂O₂
MDL Number:MFCD00988254
:128.1300 g/mol
EINECS:226-388-3
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:182.0 to 186.0 deg-C
Specification Test Results
Frequently Bought Together