


1,4-Phenylene diisocyanate ≥98.0% (by GC)
: TCI
:104-49-4
:
:C₈H₄N₂O₂
:118°C
:Refrigerator
:203-207-6
:2811
:6.1,III
:GHS05
:GHS06
:GHS08
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:94.0 to 98.0 deg-C
Frequently Bought Together