Diethyl dipropylmalonate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6065-63-0
:
Formula:C₁₃H₂₄O₄
MDL Number:MFCD00026840
:244.33 g/mol
Boiling Pt:249°C
EINECS:227-998-2
Specification Test Results
- Appearance:Colorless to Orange to Green clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together