Order Entry
Italy
ContactUsLinkComponent
Diethyl dibutylmalonate ≥98.0% (by GC)
Diethyl dibutylmalonate ≥98.0% (by GC)
  TCIAD1184-25ML
 :  TCI
 :  
undefined
Diethyl dibutylmalonate ≥98.0% (by GC)
  TCIAD1184-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :142°C
 :272.39 g/mol
 :209-890-7
 :596-75-8
 :C₁₅H₂₈O₄
 :MFCD00026842