

1,3-Dicyclohexylimidazolium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:286014-38-8
:
Formula:C₁₅H₂₅BF₄N₂
MDL Number:MFCD04973310
:320.18 g/mol
UN:1759
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:172.0 to 176.0 deg-C
Specification Test Results
Frequently Bought Together