Order Entry
Italy
ContactUsLinkComponent
2,6-Pyridinedimethanol ≥98.0% (by GC, titration analysis)
Irritante
2,6-Pyridinedimethanol ≥98.0% (by GC, titration analysis)
Nome del fornitore:  TCI
Certificati
undefined
Irritante
2,6-Pyridinedimethanol ≥98.0% (by GC, titration analysis)
Nome del fornitore:  TCI

Risultati dei test sulle specifiche

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    112.0 to 115.0 deg-C
  • Solubility in Water:
    almost transparency

Risultati dei test sulle specifiche

Frequently Bought Together