
N-Ethylpyridine-4-methylamine ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:33403-97-3
:
Formula:C₈H₁₂N₂
MDL Number:MFCD00023632
:136.20 g/mol
Flash Pt:98°C
EINECS:251-506-5
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together