
Naphthalene-2,7-dicarbonitrile ≥94.0% (by GC)
Catalog # TCIAD3705-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 94.0 %
Specification Test Results
About this item
:
Melting Pt:267°C
UN:3439
ADR:6.1,III
:GHS06
CAS Number:39718-11-1
Formula:C₁₂H₆N₂