Order Entry
Italy
ContactUsLinkComponent
1-Acetyl-2-phenylhydrazine ≥98.0% (by GC)
Toxic
1-Acetyl-2-phenylhydrazine ≥98.0% (by GC)
Catalog # TCIAA0107-25G
Supplier:  TCI
CAS Number:  
undefined
Toxic
1-Acetyl-2-phenylhydrazine ≥98.0% (by GC)
Catalog # TCIAA0107-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    129.0 to 131.0 deg-C
  • Solubility in Methanol:
    very faint turbidity

Specification Test Results

About this item

 :
 :150.18 g/mol
EINECS:204-055-3
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:114-83-0
Formula:C₈H₁₀N₂O
MDL Number:MFCD00008672