2-Trimethylsilyl-1,3-dithiane ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:13411-42-2
:
Formula:C₇H₁₆S₂Si
MDL Number:MFCD00006655
:192.417 g/mol
Boiling Pt:77°C
Flash Pt:96°C
EINECS:236-504-4
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 96.0 %
- Specific gravity (20/20):1.0380 to 1.0410
- Refractive index n20/D:1.5310 to 1.5340
Specification Test Results
Frequently Bought Together