
2,4-Hexadiyne-1,6-diol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3031-68-3
:
Formula:C₆H₆O₂
MDL Number:MFCD00020619
:110.11 g/mol
Storage Temperature:Refrigerator
EINECS:221-210-0
UN:1325
ADR:4.1,III
:GHS02
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:112.0 to 115.0 deg-C
Specification Test Results
Frequently Bought Together