
Succinic acid dihydrazide ≥97.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4146-43-4
:
Formula:C₄H₁₀N₄O₂
MDL Number:MFCD00007613
:146.1478 g/mol
Melting Pt:168°C
EINECS:223-970-9
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together