Sudan black B
TCIAS0113-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
- Appearance:Dark green to Dark red to Black powder to crystal
- Lambda max.:590.0 to 610.0 nm (EtOH)
- Absorbance (E1%1cm):min. 630 (EtOH, 590.0 to 610.0 nm)
: CI 26150, Ceres black BN, Solvent Black 3, Fat Black HB
:
:456.55 g/mol
:124°C
:Ambient
:224-087-1
:4197-25-5
:C₂₉H₂₄N₆
:MFCD00006919