
9,10-Bis(phenylethynyl)anthracene ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:10075-85-1
:
Formula:C₃₀H₁₈
MDL Number:MFCD00012050
:378.47 g/mol
Merck Index:14,01299
EINECS:233-210-8
:GHS07
Specification Test Results
- Appearance:Orange to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Chemiluminescence test:to pass test
Specification Test Results
Frequently Bought Together