
(1S)-(-)-Camphanic chloride ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:39637-74-6
:
Formula:C₁₀H₁₃ClO₃
MDL Number:MFCD00135626
:216.67 g/mol
Storage Temperature:Refrigerator
EINECS:254-552-4
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Argentmetric Titration):min. 97.0 %
- Melting point:68.0 to 72.0 deg-C
- Solubility in Toluene:almost transparency
Specification Test Results
Related Information
Frequently Bought Together