Order Entry
Italy
ContactUsLinkComponent
2,4-Dichloro-6,7-dimethoxyquinazoline ≥98.0% (by HPLC, total nitrogen)
Irritant
2,4-Dichloro-6,7-dimethoxyquinazoline ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAD3746-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2,4-Dichloro-6,7-dimethoxyquinazoline ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAD3746-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %

Specification Test Results

About this item

 :
 :259.0887 g/mol
 :GHS07
CAS Number:27631-29-4
Formula:C₁₀H₈Cl₂N₂O₂
MDL Number:MFCD00051733