Order Entry
Italy
ContactUsLinkComponent
4,6-Diaminopyrimidine-2-thiol ≥95.0% (by titrimetric analysis)
4,6-Diaminopyrimidine-2-thiol ≥95.0% (by titrimetric analysis)
Catalog # TCIAD2063-25G
Supplier:  TCI
CAS Number:  
4,6-Diaminopyrimidine-2-thiol ≥95.0% (by titrimetric analysis)
Catalog # TCIAD2063-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (Nonaqueous Titration):
    min. 95.0 %
  • Solubility in hot Acetic acid:
    almost transparency

Specification Test Results

About this item

 :
 :142.18 g/mol
EINECS:213-721-2
CAS Number:1004-39-3
Formula:C₄H₆N₄S
MDL Number:MFCD00006078