Order Entry
Italy
ContactUsLinkComponent
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) ≥98.0% (by titrimetric analysis)
Irritant
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Iodometric Titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together