Order Entry
Italy
ContactUsLinkComponent
3,3-Dimethylglutarimide ≥95.0% (by total nitrogen basis)
Irritant
3,3-Dimethylglutarimide ≥95.0% (by total nitrogen basis)
Catalog # TCIAD1324-10G
Supplier:  TCI
CAS Number:  
Irritant
3,3-Dimethylglutarimide ≥95.0% (by total nitrogen basis)
Catalog # TCIAD1324-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 95.0 %
  • Melting point:
    145.0 to 148.0 deg-C

Specification Test Results

About this item

 :
 :141.17 g/mol
EINECS:214-373-4
 :GHS07
CAS Number:1123-40-6
Formula:C₇H₁₁NO₂
MDL Number:MFCD00006671