
2,4'-Bipyridyl ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:581-47-5
:
Formula:C₁₀H₈N₂
MDL Number:MFCD00006217
:156.19 g/mol
Boiling Pt:110°C
EINECS:209-467-7
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:59.0 to 64.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together