
10-Aminodecan-1-ol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:23160-46-5
:
Formula:C₁₀H₂₃NO
MDL Number:MFCD02094313
:173.2957 g/mol
Boiling Pt:154°C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:71.0 to 75.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together