

(±)-2-Methyltetrahydrofuran-3-one ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3188-00-9
:
Formula:C₅H₈O₂
MDL Number:MFCD00010423
:100.12 g/mol
Boiling Pt:139°C
Flash Pt:38°C
EINECS:221-685-4
UN:1224
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together