

1,1,2,2-Tetrachloro-1,2-dimethyldisilane ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4518-98-3
:
Formula:C₂H₆Cl₄Si₂
MDL Number:MFCD00192469
Flash Pt:49°C
Storage Temperature:Refrigerator
EINECS:224-844-6
UN:2988
ADR:4.3,I
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together