
Tetracycline hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:64-75-5
:
Formula:C₂₂H₂₅ClN₂O₈
MDL Number:MFCD00078142
:480.9 g/mol
Merck Index:14,09196
Melting Pt:214°C
EINECS:200-593-8
:GHS08
Storage Temperature:Freezer
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:-240 to -255 deg (C=0.5, 0.1mol/L HCl)
Specification Test Results
Frequently Bought Together