Vitamin K1 (phytomenadione, phylloquinone) ≥97.0% (by UV/Vis)
Catalog # TCIAP0642-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Purity (HPLC):min. 92.0 area%
- Purity (UV-vis method):min. 97.0 %
- abs-ratio (a/b):0.69 to 0.73 (E253.5/E248.5,Isooctane sol. (1 to 100000))
- abs-ratio (2a/2b):0.28 to 0.34 (E284.5/E326.0,Isooctane sol. (1 to 10000))
Specification Test Results
About this item
:
:450.6958 g/mol
Merck Index:14,07380
Storage Temperature:Refrigerator
EINECS:201-564-2
CAS Number:84-80-0
:81818-54-4
Formula:C₃₁H₄₆O₂
MDL Number:MFCD00214063