5-Chloro-3-methylbenzo[b]thiophene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:19404-18-3
:
Formula:C₉H₇ClS
MDL Number:MFCD00052506
:182.67 g/mol
Boiling Pt:144°C
Specification Test Results
- Appearance:White to Yellow to Orange powder to lump
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together